Learn More
(R)-5-Methylhydantoin (app 80% ee), TRC
Specifications
Specifications
| Chemical Name or Material | (R)-5-Methylhydantoin |
| CAS | 55147-68-7 |
| Molecular Formula | C4 H6 N2 O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C4H6N2O2/c1-2-3(7)6-4(8)5-2/h2H,1H3,(H2,5,6,7,8)/t2-/m1/s1 |
| SMILES | C[C@H]1NC(=O)NC1=O |
| IUPAC Name | (5R)-5-methylimidazolidine-2,4-dione |
| Molecular Weight (g/mol) | 114.1 |
| Formula Weight | 114.0429 |