missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-4-Fluorostyrene oxide, 98+%, ee 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications
Density | 1.17 |
Boiling Point | 92°C (14 mmHg) |
Refractive Index | 1.5087 |
Quantity | 1 g |
UN Number | UN1992 |
Solubility Information | Sparingly soluble in water.(0.26g/L) (25°C), |
Formula Weight | 138.14 |
Percent Purity | ≥98% |
Assay | ee ≥98% |
Chemical Name or Material | (R)-(-)-4-Fluorostyrene oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'