Learn More
(R)-4-Benzylmorpholine-2-carbonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-4-Benzylmorpholine-2-carbonitrile |
| CAS | 2199389-10-9 |
| Molecular Formula | C12H14N2O |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C12H14N2O/c13-8-12-10-14(6-7-15-12)9-11-4-2-1-3-5-11/h1-5,12H,6-7,9-10H2/t12-/m0/s1 |
| SMILES | N#C[C@H]1CN(Cc2ccccc2)CCO1 |
| IUPAC Name | (2R)-4-benzylmorpholine-2-carbonitrile |
| Molecular Weight (g/mol) | 202.25 |
| Formula Weight | 202.1106 |