Learn More
(R)-(-)-3-(Carbamoylmethyl)-5-methylhexanoic acid, TRC
Product Code. 30137729
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137729 1 g 1g
30137717 5 g 5g
30011112 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137729 Supplier Toronto Research Chemicals Supplier No. TRCC1759551G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 181289-33-8
Molecular Formula C9 H17 N O3
Molecular Weight (g/mol) 187.24
Synonym (3R)-3-(2-amino-2-oxoethyl)-5-methylhexanoic acid, Hexanoic acid, 3-(2-amino-2-oxoethyl)-5-methyl-, (R)-, (3R)-3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid, (R)-(-)-3-(Carbamoylmethyl)-5-methylhexanoic acid, (R)-3-(Carbamoylmethyl)-5-methylhexanoic acid
IUPAC Name (3R)-3-(2-amino-2-oxoethyl)-5-methylhexanoic acid
SMILES CC(C)C[C@H](CC(=O)N)CC(=O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H17NO3/c1-6(2)3-7(4-8(10)11)5-9(12)13/h6-7H,3-5H2,1-2H3,(H2,10,11)(H,12,13)/t7-/m1/s1
Formula Weight 187.1208
Percent Purity >95%
Chemical Name or Material (R)-3-(Carbamoylmethyl)-5-methylhexanoic Acid
Grade HPLC