Learn More
(R)-3-(Boc-amino)piperidine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | (R)-3-(Boc-amino)piperidine |
| CAS | 309956-78-3 |
| Molecular Formula | C10H20N2O2 |
| Quantity | 5 g |
| Synonym | (3R)-3-Piperidinyl-carbamic Acid 1,1-Dimethylethyl Ester, (3R)-3-[((tert-Butyloxycarbonyl)amino)]piperidine, (R)-3-(tert-Butoxycarbonylamino)piperidine, (R)-3-[(tert-Butoxycarbonyl)amino]piperidine, (R)-3-[(tert-Butyloxycarbonyl)amino]piperidine, (R)-3-[N-(tert-Butoxycarbonyl)amino]piperidine, (R)-3-tert-Butoxycarbonylaminopiperidine, (R)-Piperidin-3-ylcarbamic acid tert-butyl ester, (R)-tert-Butyl (Piperidin-3-yl)carbamate, tert-Butyl (R)-N-(3-Piperidyl)carbamate, tert-Butyl (R)-N-(Piperidin-3-yl)carbamate, tert-Butyl (R)-piperidin-3-ylcarbamate |
| InChI Formula | InChI=1S/C10H20N2O2/c1-10(2,3)14-9(13)12-8-5-4-6-11-7-8/h8,11H,4-7H2,1-3H3,(H,12,13)/t8-/m1/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@@H]1CCCNC1 |
| IUPAC Name | tert-butyl N-[(3R)-piperidin-3-yl]carbamate |
| Molecular Weight (g/mol) | 200.28 |
| Formula Weight | 200.1525 |