Learn More
(R)-2-Phenyl-2-pivalamidoacetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-phenylacetic Acid |
| CAS | 40610-41-1 |
| Molecular Formula | C13 H17 N O3 |
| Quantity | 100 mg |
| Synonym | (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-phenylacetic Acid, Ampicillin Sodium Imp. K (EP) |
| InChI Formula | InChI=1S/C13H17NO3/c1-13(2,3)12(17)14-10(11(15)16)9-7-5-4-6-8-9/h4-8,10H,1-3H3,(H,14,17)(H,15,16)/t10-/m1/s1 |
| SMILES | CC(C)(C)C(=O)N[C@@H](C(=O)O)c1ccccc1 |
| IUPAC Name | (2R)-2-(2,2-dimethylpropanoylamino)-2-phenylacetic acid |
| Molecular Weight (g/mol) | 235.28 |
| Formula Weight | 235.1208 |