Learn More
(R)-2-Methylbutyric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-2-Methylbutyric Acid |
| CAS | 32231-50-8 |
| Molecular Formula | C5H10O2 |
| Quantity | 1 g |
| Synonym | (2R)-2-Methylbutanoic Acid, (-)-2-Methylbutanoic Acid, (R)-(-)-2-Methylbutanoic Acid |
| InChI Formula | InChI=1S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)/t4-/m1/s1 |
| SMILES | CC[C@@H](C)C(=O)O |
| IUPAC Name | (2R)-2-methylbutanoic acid |
| Molecular Weight (g/mol) | 102.13 |
| Formula Weight | 102.0681 |