missing translation for 'onlineSavingsMsg'
Learn More

(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%

Product Code. p-7040354
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495445 250 mg 250mg
15485445 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495445 Supplier Thermo Scientific Alfa Aesar Supplier No. H60112.MD
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1365531-98-1 | C76H108O8P2 | 1211.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Incompatible with light and oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1365531-98-1
Molecular Formula C76H108O8P2
Molecular Weight (g/mol) 1211.64
MDL Number MFCD20265636
InChI Key KACYLFSRRUJDSY-UHFFFAOYSA-N
Synonym r-dtbm-garphos, 4,4',6,6'-tetramethoxy-1,1'-biphenyl-2,2'-diyl bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphine, ar-2,2'-bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino-4,4'-dimethoxy-6,6'-dimethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphine, r-2,2'-bis bis 4-methoxy-3,5-di-t-butylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, r-2,2'-bis bis 4-methoxy-3,5-di-t-butylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphine, s-2,2'-bis bis 4-methoxy-3,5-di-t-butylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, 2'-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 3,5-di-tert-butyl-4-methoxyphenyl phosphane
PubChem CID 87110675
IUPAC Name [2-[2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane
SMILES CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC(=C(C(=C2)C(C)(C)C)OC)C(C)(C)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=C(C(=C5)C(C)(C)C)OC)C(C)(C)C)C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)OC)OC)OC)OC

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 1211.61
Percent Purity ≥97%
Chemical Name or Material (R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.