Learn More
(R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, TRC
Specifications
Specifications
| Chemical Name or Material | (1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline |
| CAS | 180272-45-1 |
| Molecular Formula | C15 H15 N |
| Quantity | 5 g |
| Synonym | Isoquinoline','1,2,3,4-tetrahydro-1-phenyl-','(1R)-, Isoquinoline','1,2,3,4-tetrahydro-1-phenyl-','(R)-, (-)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (1R)-Phenyl-1,2,3,4-tetrahydroisoquinoline, (R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline |
| InChI Formula | InChI=1S/C15H15N/c1-2-7-13(8-3-1)15-14-9-5-4-6-12(14)10-11-16-15/h1-9,15-16H,10-11H2/t15-/m1/s1 |
| SMILES | C1Cc2ccccc2[C@H](N1)c3ccccc3 |
| IUPAC Name | (1R)-1-phenyl-1,2,3,4-tetrahydroisoquinoline |
| Molecular Weight (g/mol) | 209.29 |
| Formula Weight | 209.1204 |