Learn More
(R)-(+)-1-Octyn-3-ol, 98+%, Thermo Scientific Chemicals
Product Code. 11349585
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349585 1 g 1g
11359585 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11349585 Supplier Thermo Scientific Alfa Aesar Supplier No. L19045.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
(R)-(+)-1-Octyn-3-ol is used as an intermediate for the synthesis of prostaglandins, modified eicosanoids structured with fatty acids attached to a 5 membered ring.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(+)-1-Octyn-3-ol is used as an intermediate for the synthesis of prostaglandins, modified eicosanoids structured with fatty acids attached to a 5 membered ring.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32556-70-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00192370
InChI Key VUGRNZHKYVHZSN-QMMMGPOBSA-N
Synonym r-+-1-octyn-3-ol, 1-octyn-3-ol, 3r, 3r-oct-1-yn-3-ol, r-1-octyn-3-ol, unii-az9v2w5sw4, 3r-1-octyn-3-ol, az9v2w5sw4, +-1-octyn-3-ol, 1-octyn-3-ol, r, 1-octyn-3-ol, +
PubChem CID 6999692
IUPAC Name (3R)-oct-1-yn-3-ol
SMILES CCCCCC(C#C)O
Specifications
Density 0.866
Boiling Point 100°C (20 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.442
Quantity 1 g
Beilstein 2037704
Optical Rotation +9.9° (Neat)
Formula Weight 126.2
Percent Purity ≥98%
Chemical Name or Material (R)-(+)-1-Octyn-3-ol
RUO – Research Use Only
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.