Learn More
(R)-1-(3-Pyridyl)ethanol, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-1-(3-Pyridyl)ethanol |
| CAS | 7606-26-0 |
| Molecular Formula | C7 H9 N O |
| Quantity | 500 mg |
| Synonym | 3-Pyridinemethanol','α-methyl-','(R)-, 3-Pyridinemethanol','α-methyl-','(R)-(+)- (8CI), (αR)-α-Methyl-3-pyridinemethanol, (1R)-1-(Pyridin-3-yl)ethan-1-ol, (1R)-1-Pyridin-3-ylethanol, (R)-(+)-1-(3-Pyridyl)ethanol, (R)-(+)-1-(Pyridin-3-yl)ethanol, (R)-1-(3-Pyridyl)ethanol, (R)-1-(3'-Pyridyl)ethanol, 3-Pyridinemethanol α-methyl-','(αR)- |
| InChI Formula | InChI=1S/C7H9NO/c1-6(9)7-3-2-4-8-5-7/h2-6,9H,1H3/t6-/m1/s1 |
| SMILES | C[C@@H](O)c1cccnc1 |
| IUPAC Name | (1R)-1-pyridin-3-ylethanol |
| Molecular Weight (g/mol) | 123.153 |
| Formula Weight | 123.068 |