missing translation for 'onlineSavingsMsg'
Learn More

QX77, MedChemExpress

Product Code. 16454638
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16454638

Brand: Medchem Express HY112483

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress QX77 is a chaperone-mediated autophagy (CMA) activator and upregulates LAMP2A expression in vitro. QX77 induces Rab11 upregulation, rescues Rab11 down-regulation and trafficking deficiency in cystinotic cells. QX77 can impede self-renewal and promote differentiation of ES cells.

QX77 is a chaperone-mediated autophagy (CMA) activator and upregulates LAMP2A expression in vitro. QX77 induces Rab11 upregulation, rescues Rab11 down-regulation and trafficking deficiency in cystinotic cells. QX77 can impede self-renewal and promote differentiation of ES cells.

Specifications

Chemical Name or Material QX77
CAS 1798331-92-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H13ClN2O2
Quantity 10 mg
Solubility Information DMSO : 15 mg/mL (49.88 mM; ultrasonic and warming and heat to 60°C)
SMILES CC(NC1=CC=C(C2=NC3=CC=C(Cl)C=C3OC2)C=C1)=O
Molecular Weight (g/mol) 300.74
Formula Weight 300.74
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.