missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-6-boronic acid hydrochloride, 95%, Thermo Scientific™

Product Code. 15403917
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403917 250 mg 250mg
15493907 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15403917 Supplier Thermo Scientific Alfa Aesar Supplier No. H30380.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310403-86-1
Molecular Formula C9H9BClNO2
Molecular Weight (g/mol) 209.436
MDL Number MFCD11101398
InChI Key XEUWKJJLVYUQIS-UHFFFAOYSA-N
Synonym quinolin-6-ylboronic acid hydrochloride, quinoline-6-boronic acid hydrochloride, 6-quinolineboronic acid hydrochloride
PubChem CID 44119111
IUPAC Name quinolin-6-ylboronic acid;hydrochloride
SMILES B(C1=CC2=C(C=C1)N=CC=C2)(O)O.Cl

Specifications

Melting Point ∽220°C (decomposition)
Quantity 250 mg
Formula Weight 209.44
Percent Purity 95%
Chemical Name or Material Quinoline-6-boronic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.