Learn More
Quinine-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Quinine-d3 |
| CAS | 2734920-95-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20H21D3N2O2 |
| Quantity | 10 mg |
| Synonym | (8α,9R)-6’-(Methoxy-d3)cinchonan-9-ol, (-)-Quinine-d3, (8S,9R)-Quinine-d3, (R)-(-)-Quinine-d3, 6'-Methoxycinchonidine-d3, Chinine-d3, NSC 192949-d3 |
| InChI Formula | InChI=1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19-,20+/m0/s1/i2D3 |
| SMILES | [H][C@@]1(C=C)[C@@H]2C[C@]([C@H](O)C3=CC=NC4=CC=C(OC([2H])([2H])[2H])C=C43)([H])[N@](C1)CC2 |
| IUPAC Name | (R)-(6-(methoxy-d3)quinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol |
| Molecular Weight (g/mol) | 327.44 |
| Show More |