Learn More
Plum™ QuickFix Water Resistant Plaster Dispenser
Product Code. 10048702
Change view
Click to view available options
Quantity:
1 Dispenser, 90 Plasters
Unit Size:
Pack of 90
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10048702 1 Dispenser, 90 Plasters Pack of 90
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10048702 Supplier Plum™ Supplier No. 5501
  • ONLINE EXCLUSIVE PRICE
    52.76 €
    30.71 € / Pack of 90
    Save 22.05 €
    41% Off
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Wall-mounted plaster dispenser with 90 water resistant plasters that are particularly suitable for working environments with a high level of humidity. The plasters are made of a protective PE material, which allows the skin to breathe.
  • Gives immediate and visible access to plasters
  • Each plaster is ready for use just by pulling it from the dispenser
  • Each plaster is packed separately
  • Only one hand is needed to apply
  • Can be refilled as required
  • Easy to see when it is empty
  • Easy to stop minor bleeding, also without staining the other plasters
  • Small dimensions - easy to find room for the QuickFix dispenser
  • Includes a shelf for storing small, relevant objects
  • Can be locked so that it can only be opened with a key - may prevent unwanted waste
  • Refill Product Article Number: 5511
  • Specifications
    Depth (Metric) 32 mm
    Quantity 1 Dispenser, 90 Plasters
    Dimensions (L x W) 72 x 25 mm
    Width (Metric) 230 mm
    Height (Metric) 135 mm