Learn More
Quickfit™ Virgeux Fractionating Columns
Product Code. 15748017
Click to view available options
Standard Taper Joint:
19/26
24/29
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 15748017
Brand: Quickfit FC3/22/15
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Quickfit™ Virgeux Fractionating Columns use a combination of horizontal and angled indentations to significantly increase the surface area of the column to increase its efficiency.
  • Easy to use column
  • Vigreux type with combination of horizontal and angled indentations
  • Horizontal indentations ensure intimate vapor and liquid contact
  • Angled indentations redistribute liquid from walls to center of column
TRUSTED_SUSTAINABILITY
Specifications
Includes Side arm and screw threads
Length (Metric) 178 mm
Type Fractionating Column
Standard Taper Joint 19/26