Learn More
Quickfit™ Unbadged Joints Cones
Product Code. 12468816
Change view
Click to view available options
Cone Size:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
40/38
45/40
55/44
7/16
Thickness (Metric) Wall:
1mm
1.5mm
1.8mm
2.00mm
2mm
2.30mm
5mm
Unit Size:
Pack of 10
Pack of 20
Pack of 5
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size Thickness (Metric) Wall unitSize
12468816 14/23 1.5mm Pack of 10
12939363 10/19 1mm Pack of 10
10830251 19/26 1.8mm Pack of 10
12614911 29/32 2mm Pack of 10
12488816 24/29 1.8mm Pack of 10
12458826 7/16 1.5mm Pack of 10
10735671 40/38 2mm Pack of 5
12653012 45/40 2.30mm Pack of 5
13431829 34/35 2.00mm Pack of 10
15597760 12/21 1.5mm Pack of 50
15507770 55/44 5mm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 12468816 Supplier Quickfit Supplier No. CNB14UB
Only null left
Add to basket
Add to basket
Free from any ceramic marking
  • Avoids problem of badge charring at high temperatures which can give an untidy appearance
  • Manufactured to accuracy which surpasses requirements of ISO 383
  • Pyrex™ borosilicate glass for resistance to chemical corrosion and thermal shock
  • Cones manufactured with shank length of 100mm
TRUSTED_SUSTAINABILITY
Specifications
Type Joint Cone
Cone Size 14/23
Thickness (Metric) Wall 1.5mm