Learn More
Pyrex™ Quickfit™ Rotary Evaporator Splash Head with Sinter
Protects the condenser against contamination. Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter prevents splash with sinter.
Brand:  Pyrex™ SH16/19
Product Code. 12914075
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • For use with rotary evaporators
  • Prevents possible contamination of the condenser from 'bumping' solvents
  • Features a coarse porosity 1 sinter to reduce carry over
  • Splash heads feature a 29/32 socket