Learn More
Pyrex™ Quickfit™ Rotary Evaporator Splash Head with Sinter
Protects the condenser against contamination. Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter prevents splash with sinter.
Brand: Pyrex™ SH16/19
This item is not returnable.
View return policy
Description
- For use with rotary evaporators
- Prevents possible contamination of the condenser from 'bumping' solvents
- Features a coarse porosity 1 sinter to reduce carry over
- Splash heads feature a 29/32 socket