Learn More
Applied Biosystems™ QuantStudio™ Pro Block Upgrade Kit, TaqMan™ Array Card
Product Code. 16836043 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16836043 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 16836043 Supplier Applied Biosystems™ Supplier No. A45956
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For Research Use Only. All usage must comply with product instructions.
The QuantStudio Pro Block Upgrade Kit is an optional accessory for the QuantStudio Pro Real-Time qPCR System.
The QuantStudio Pro Block Upgrade Kit is an optional accessory for the QuantStudio Pro Real-Time qPCR System. Interchangeable blocks enable application versatility with easy, powered-up interchange of block types.
The QuantStudio 7 Pro system accommodates the interchange of 96-well (0.2 mL), 96-well (0.1 mL), 384-well, and TaqMan array card (TAC) blocks. The system is delivered with one block type; other block types may be purchased separately. For a more value-priced system that does not support TaqMan array cards or automation (but that can be upgraded to do so at a later date), please see our QuantStudio 6 Pro Real-Time PCR System. And for the ultimate in flexibility, consider the QuantStudio 12K Flex Real-Time PCR System, which also accommodates TaqMan OpenArray plates.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) QuantStudio™ 7 Pro
Product Type Block Upgrade
Quantity 1 Kit