Learn More
Applied Biosystems™ QuantStudio™ Absolute Q™ AutoRun Digital PCR Suite, single to dual configuration upgrade
Shop All Applied Biosystems ProductsDescription
Upgrade your existing QuantStudio Absolute Q AutoRun Digital PCR Suite, Single Configuration, to the QuantStudio Absolute Q AutoRun Digital PCR Suite, Dual Configuration, an integrated system solution that scales precise nucleic acid quantification and offers intuitive automation to help support productivity and accelerate progress.
The QuantStudio Absolute Q AutoRun Digital PCR Suite, Dual Configuration, consists of two QuantStudio Absolute Q Digital PCR systems integrated with the Spinnaker Microplate Mover via the easy-to-use Momentum Workflow Scheduler Software to manage loading and unloading of plates for an automated workflow. By harnessing the power of two digital PCR systems in your automated workflow, you can double your throughput while fortifying the resilience of your operations.
This upgrade includes a location bracket, a nest adapter, necessary cables, and an updated Thermo Scientific™ Momentum Workflow Software license to support integration of the second QuantStudio Absolute Q Digital PCR System.
Specifications
Specifications
| For Use With (Equipment) | Absolute Q™ |
| Voltage | 100/240 V |
| For Use With (Application) | Digital PCR |
| Quantity | 1 upgrade |
| Calibration Type | Positional Teaching |