missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Quanti-Cult™ Staphylococcus aureus subsp. aureus ATCC™ 25923™

Product Code. 12627699
Click to view available options
Quantity:
10 Tests/Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12627699

Brand: Thermo Scientific™ R4737010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.

Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media that can be stored at room-temperature to free up incubator space.

  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum

Ordering Alerts:

Yiels 1 Test/reconstituted vial, 10 Tests/kit

Includes:

10 preserved organism vials and 10 rehydration vials.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 25923TM*
Description Staphylococcus aureus subsp. aureus ATCC™ 25923™
Format Quanti-Cult
Product Type Bactidisk
Target Organism Class Cocci
Test Type Quantitative QC
Packaging Type 10 Test/Kit
Quantity 10 Tests/Kit
Strain Designations Seattle 1944

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.