Learn More
Socorex™ Qualitix™ Tipfill Refill System
Product Code. 15760649
Change view
Click to view available options
Tip Style:
Beveled
Fine Point
Unit Size:
Pack of 480
Pack of 960
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tip Style unitSize
15760649 Beveled Pack of 960
15750659 Fine Point Pack of 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15760649 Supplier Socorex™ Supplier No. 3070.0200T
Only null left
Add to basket
Add to basket
Intended for easy rack loading. Socorex™ Qualitix™ Tipfill Refill System are filled trays for instant transfer into a rack without risk of contamination.
  • Easy rack loading.
  • Instant transfer without contamination risk.
  • Smart, re-closable Tipfill box reduces storage space, waste volume, and shipping costs.
Specifications
Volume (Metric) 200 μL
Volume (Metric) Dispensing 200μL
Color Natural
Tip Style Beveled
Packaging 96/Rack
Quantity 10/PK, 10PK/CS
Disposable Single-use
Product Line Qualitix