Learn More
Hellma™ QS Quartz Suprasil High Precision Macro Fluorescence Cells With Polyethylene Stopper
Product Code. 11731854
Click to view available options
Capacity (Metric):
3500 μL
Unit Size:
Each
Product Code. 11731854
Brand: Hellma™ 1011020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Macro Fluorescence Cell
Capacity (Metric) 3500 μL
Material Optical Glass
Width (Metric) Interior 10mm
Path Length 10 x 10 mm
Height (Metric) 45 mm
Width (Metric) 12.5 mm