Learn More
Izon qEVsingle Starter Pack with qEV Rack and 40 qEVsingle Columns
Specifications
Specifications
| Description | Izon qEVsingle stater pack with qEV rack and 40 qEVsingle columns |
| Includes | qEV rack and 40 qEVsingle columns |
| Column Type | Size Exclusion Column |
| For Use With (Application) | EV/exsosome isolation |