Learn More
Izon qEV2 Column
Product Code. 15948090
Encompass_Preferred
Change view
Click to view available options
For use with EV/exsosome isolation
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15948090 For use with EV/exsosome isolation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15948090 Supplier Izon Supplier No. SP4
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
qEV provides the most rapid and effective EV/exsosome isolation system available. qEV is based on Size Exclusion Chromatography (SEC), and takes around 15 minutes to get a pure sample of intact exosomes. The resulting samples are consistent, standardisable and repeatable.
  • The qEV2 is designed for separating exosomes from a 2mL biological sample and comes with Luer Lock fittings. They will be of particular interest if you are working with larger fluid volumes. The Luer Lock fittings allow for many different configurations
Specifications
Description Izon qEV2
Column Type Size Exclusion Column
For Use With (Application) EV/exsosome isolation