missing translation for 'onlineSavingsMsg'
Learn More

PZM21, MedChemExpress

Product Code. 16410759
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16410759 1 mg 1mg
16486067 5 mg 5mg
16474586 100 mg 100mg
16469876 50 mg 50mg
16413348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16410759 Supplier Medchem Express Supplier No. HY101386

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress PZM21 is a potent and selective μ opioid receptor agonist with an EC50 of 1.8 nM.

PZM21 is a potent and selective μ opioid receptor agonist with an EC50 of 1.8 nM.

Specifications

Chemical Name or Material PZM21
CAS 1997387-43-5
Color White
Purity Grade Notes Research
Molecular Formula C19H27N3O2S
Quantity 1 mg
Solubility Information DMSO : 100 mg/mL (276.63 mM; Need ultrasonic)
SMILES O=C(N[C@@H](C)CC1=CSC=C1)NC[C@@H](N(C)C)CC2=CC=C(O)C=C2
Molecular Weight (g/mol) 361.5
Formula Weight 361.5
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.