Learn More
PYZD-4409, MedChemExpress
Product Code. 16475628
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475628 10 mg 10mg
16466156 100 mg 100mg
16451797 50 mg 50mg
16428327 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16475628 Supplier Medchem Express Supplier No. HY13297
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress PYZD-4409 is a specific inhibitor of the ubiquitin-activating enzyme UBA1 with an IC50 of 20 μM (cell-free enzymatic assay). PYZD-4409 induces cell death in malignant cells and preferentially inhibits the clonogenic growth of primary acute myeloid leukemia cells.
PYZD-4409 is a specific inhibitor of the ubiquitin-activating enzyme UBA1 with an IC50 of 20 μM (cell-free enzymatic assay). PYZD-4409 induces cell death in malignant cells and preferentially inhibits the clonogenic growth of primary acute myeloid leukemia cells.
Specifications
Chemical Name or Material PYZD-4409
CAS 423148-78-1
Color Light Brown to Black
Purity Grade Notes Research
Molecular Formula C14H7ClFN3O5
Quantity 10 mg
Solubility Information DMSO : ≥ 35 mg/mL (99.53 mM)
SMILES FC1=CC=C(N2C(/C(C(N2)=O)=C\C3=CC=C([N+]([O-])=O)O3)=O)C=C1Cl
Molecular Weight (g/mol) 351.67
Formula Weight 351.67
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease