missing translation for 'onlineSavingsMsg'
Learn More

PYZD-4409, MedChemExpress

Product Code. 16466156
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466156 100 mg 100mg
16475628 10 mg 10mg
16451797 50 mg 50mg
16428327 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466156 Supplier Medchem Express Supplier No. HY13297
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PYZD-4409 is a specific inhibitor of the ubiquitin-activating enzyme UBA1 with an IC50 of 20 μM (cell-free enzymatic assay). PYZD-4409 induces cell death in malignant cells and preferentially inhibits the clonogenic growth of primary acute myeloid leukemia cells.

PYZD-4409 is a specific inhibitor of the ubiquitin-activating enzyme UBA1 with an IC50 of 20 μM (cell-free enzymatic assay). PYZD-4409 induces cell death in malignant cells and preferentially inhibits the clonogenic growth of primary acute myeloid leukemia cells.

Specifications

Chemical Name or Material PYZD-4409
CAS 423148-78-1
Color Light Brown to Black
Purity Grade Notes Research
Molecular Formula C14H7ClFN3O5
Quantity 100 mg
Solubility Information DMSO : ≥ 35 mg/mL (99.53 mM)
SMILES FC1=CC=C(N2C(/C(C(N2)=O)=C\C3=CC=C([N+]([O-])=O)O3)=O)C=C1Cl
Molecular Weight (g/mol) 351.67
Formula Weight 351.67
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.