Learn More
Pyruvic acid sodium salt, Millipore™
Description
Zwitterionic buffer substance MES is used as a zwitterionic buffer substance in the preparation of media for the culture of microorganisms and plant cells. MES is a very convenient buffer substance due to its biological inertness.
Specifications
Specifications
| Chemical Name or Material | Pyruvic acid sodium salt |
| Melting Point | 236.8°C |
| CAS | 113-24-6 |
| Quantity | 50 g |
| Density | 1.78 g/cm³ at 20°C |
| Synonym | Pyruvic acid sodium salt, Sodium pyruvate, Pyruvate, Pyruvic acid sodium salt |
| Molecular Weight (g/mol) | 110.04 |
| Grade | Not Available |