Learn More
Pyrilamine Maleate Salt, TRC
Product Code. 30180389
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180389 5 g 5g
30179801 1 g 1g
30180029 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30180389 Supplier Toronto Research Chemicals Supplier No. TRCP9972255G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59-33-6
Molecular Formula C17 H23 N3 O . C4 H4 O4
Molecular Weight (g/mol) 401.46
Synonym 1,2-Ethanediamine, N-[(4-methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-, (2Z)-2-butenedioate (1:1) (9CI), 1,2-Ethanediamine, N-[(4-methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-, (Z)-2-butenedioate (1:1), Pyridine, 2-[[2-(dimethylamino)ethyl](p-methoxybenzyl)amino]-, maleate (1:1) (8CI), 2-[(2-Dimethylaminoethyl)(p-methoxybenzyl)amino]-pyridine maleate, 2-[[2-(Dimethylamino)ethyl](p-methoxybenzyl)amino]pyridine Bimaleate, AH, Anisopyradamine, Antamine, Anthisan maleate, Antihist, Antisan, Diaminide maleate, Enrumay, Histalet Forte, Histalon, Histan, Histapyran, Histatex, Histavet P, Mepyramine hydrogen maleinate, Mepyramine maleate, Minihist, N-p-Methoxybenzyl-N',N'-dimethyl-N-α-pyridylethylenediamine maleate, NSC 3604, Neo-Antergan maleate, Neoantergan maleate, Paramal, Paraminyl, Paraminyl maleate, Parmal, Prefrin A, Pymafed, Pyra Maleate, Pyramal, Pyranilamine maleate, Pyraninyl, Pyranisamine maleate, Pyrilamine maleate, Renstamin, Stamine, Stangen, Stangen maleate, Statomin maleate, Thylogen, Thylogen maleate
IUPAC Name (Z)-but-2-enedioic acid;N'-[(4-methoxyphenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine
SMILES COc1ccc(CN(CCN(C)C)c2ccccn2)cc1.OC(=O)\C=C/C(=O)O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C17H23N3O.C4H4O4/c1-19(2)12-13-20(17-6-4-5-11-18-17)14-15-7-9-16(21-3)10-8-15;5-3(6)1-2-4(7)8/h4-11H,12-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formula Weight 401.1951
Chemical Name or Material Mepyramine maleate