missing translation for 'onlineSavingsMsg'
Learn More

Pyridostatin hydrochloride, MedChemExpress

Product Code. p-200101188
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16472457

Brand: Medchem Express HY15176A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyridostatin (RR82) hydrochloride is a G-quadruplex DNA stabilizing agent (Kd=490 nM). Pyridostatin hydrochloride promotes growth arrest in human cancer cells by inducing replication- and transcription-dependent DNA damage. Pyridostatin hydrochloride targets the proto-oncogene Src. Pyridostatin hydrochloride reduced SRC protein levels and SRC-dependent cellular motility in human breast cancer cells.

Pyridostatin (RR82) hydrochloride is a G-quadruplex DNA stabilizing agent (Kd=490 nM). Pyridostatin hydrochloride promotes growth arrest in human cancer cells by inducing replication- and transcription-dependent DNA damage. Pyridostatin hydrochloride targets the proto-oncogene Src. Pyridostatin hydrochloride reduced SRC protein levels and SRC-dependent cellular motility in human breast cancer cells.

Specifications

Chemical Name or Material Pyridostatin hydrochloride
CAS 1781882-65-2
Color White
Purity Grade Notes Research
Molecular Formula C31H37Cl5N8O5
Quantity 50 mg
Synonym RR82 hydrochloride
Solubility Information H2O : 50 mg/mL (64.19 mM; Need ultrasonic)
SMILES O=C(C1=CC(OCCN)=CC(C(NC2=NC3=C(C(OCCN)=C2)C=CC=C3)=O)=N1)NC4=CC(OCCN)=C5C=CC=CC5=N4.Cl.Cl.Cl.Cl.Cl
Molecular Weight (g/mol) 778.94
Formula Weight 778.94
Percent Purity 98.77%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.