Learn More
Pyrex™ Polypropylene Luer Fittings
Description
- Full material traceability
- Polypropylene
- Female luer
- 11/64 in. Hose barbs
Specifications
Specifications
| Certifications/Compliance | ISO 594-1, Manufactured from 100% virgin raw materials in Class 100,000 environment |
| Material | Polypropylene |
| Description | Female luer to 0.17 in. low profile semi-rigid tubing barb |
| For Use With (Equipment) | Syringe |
| Connection Type | Port 1 - female luer, port 2 - 11/64 in. hose barb |