Learn More
Pyrex™ PBT Screwcaps
Click to view available options
Thread Size (Metric):
13 mm
15 mm
20 mm
22 mm
25 mm
30 mm
42 mm
Unit Size:
Pack of 10
Pack of 20
Description
- PTFE faced sealing disc for chemical resistance
- Withstands hot air sterilization to 180deg.C
- Autoclavable
Specifications
Specifications
| Description | Screw cap |
| Includes | PTFE-faced Rubber Insert |
| Material | PBT |
| Quantity | 20 Pk. |
| Thread Size (Metric) | 15 mm |
| Liner Material | PTFE-faced Rubber |
| Product Type | Screw Cap |
| For Use With (Equipment) | Culture tubes |
| Autoclavable | Autoclavable |