Learn More
Pyrex™ Centrifuge Tubes with Conical Bottom
Description
- Manufactured from chemically inert Pyrex™ glass for contamination free performance
- Conical shape with rim
- Can be centrifuged up to 3, 000 RCF values
Specifications
Specifications
| Color | Clear |
| Capacity (Metric) | 12 mL |
| Material | Borosilicate Glass |
| No. per Pack | 40 |
| Length (Metric) | 110 mm |
| Bottom Shape | Conical |
| Diameter (Metric) | 17 mm |
| Graduated | No |
| Max. RCF | 3000 x g |