Learn More
Pyrex™ Borosilicate Glass Non-vacuum Desiccator Cover
Click to view available options
Diameter (Metric) Inner:
Flange: 170 mm
Flange: 220 mm
Unit Size:
Each
Description
Made of Borosilicate glass
Spare cover for non-vacuum desiccator
Specifications
Specifications
| No. per Pack | Each |
| Product Type | Spare Cover |
| Material | Borosilicate Glass |
| For Use With (Application) | for drying under atmospheric pressure |
| For Use With (Equipment) | Non-vacuum desiccator |
| Diameter (Metric) Inner | Flange: 220 mm |