missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ 40mm Diameter Glass Büchner Filter Funnel
Description
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
- Choice of disc diameters and porosities
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Diameter (Metric) Stem | 8mm |
| Material | Borosilicate Glass |
| Height (Metric) | 170 mm |
| For Use With (Application) | For convenient filtration of solutions |
| Diameter (Metric) Disc | 40mm |
| Includes | Integrated filter plate sintered Glass disc |
| Length (Metric) Stem | 100mm |
| Porosity | 1 |
| Pore Size | 100 to 160μm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?