Learn More
Pyrex™120mm Diameter Glass Büchner Filter Funnel
Manufactured from chemically resistant Pyrex™ glass. Pyrex™ 120mm Diameter Glass Büchner Filter Funnel with integral sintered glass disc is highly resistant to most corrosive reagents and eliminates the need for filter papers.
Brand: Pyrex 3720/08M
498.75 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 1.36800kg
Description
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid - Choice of disc diameters and porosities
Specifications
1000 mL | |
Pyrex Glass, Borosilicate Glass | |
For easy filtering solutions | |
Integrated filter plate sintered Glass disc | |
4 | |
Buchner filter funnel |
19mm | |
290 mm | |
120mm | |
120mm | |
10 to 16μm |