missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinamide, 99%, MP Biomedicals™

Product Code. p-4405830 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455402 10 g 10g
11465402 25 g 25g
11475402 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455402 Supplier MP Biomedicals Supplier No. 0219547310
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 98-96-4
Molecular Formula C5H5N3O
Molecular Weight (g/mol) 123.115
InChI Key IPEHBUMCGVEMRF-UHFFFAOYSA-N
Synonym pyrazinamide, pyrazinecarboxamide, zinamide, pyrazinoic acid amide, 2-pyrazinecarboxamide, aldinamide, aldinamid, pirazinamid, pyrazineamide, pyrafat
PubChem CID 1046
ChEBI CHEBI:45285
IUPAC Name pyrazine-2-carboxamide
SMILES C1=CN=C(C=N1)C(=O)N

Specifications

Color White
Quantity 10 g
Formula Weight 123.1
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material Pyrazinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.