missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PYGM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119155
This item is not returnable.
View return policy
Description
PYGM Polyclonal Antibody for ICC/IF, IHC (P), Western Blot
This gene encodes a muscle enzyme involved in glycogenolysis. Highly similar enzymes encoded by different genes are found in liver and brain. Mutations in this gene are associated with McArdle disease (myophosphorylase deficiency), a glycogen storage disease of muscle. Alternative splicing results in multiple transcript variants.
Specifications
| PYGM | |
| Polyclonal | |
| Unconjugated | |
| PYGM | |
| AI115133; glycogen muscle; glycogen phosphorylase; glycogen phosphorylase, muscle associated; glycogen phosphorylase, muscle form; GPMM; muscle (McArdle syndrome); muscle glycogen phosphorylase; Muscpho; Myophosphorylase; PG; Phosphorylase glycogen; phosphorylase, glycogen, muscle; Phosphorylase, glycogen; muscle (McArdle syndrome); Pygm; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 19309, 24701, 5837 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P09812, P11217, Q9WUB3 | |
| PYGM | |
| Recombinant protein Glycogen Phosphorylase, Muscle. The antigen corresponds to amino acid range 11-187 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction