missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The MW of this protein is 102 kDa, and this antibody specifically recognizes the 102 kDa protein.
PWP2 belongs to the WD repeat PWP2 family. It contains 14 WD repeats. The exact function of PWP2 is not known.
Specifications
Specifications
| Antigen | PWP2 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | PWP2 |
| Gene Accession No. | Q15269 |
| Gene Alias | EHOC 17, PWP2, PWP2H |
| Gene Symbols | PWP2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?