Learn More
Invitrogen™ PWP1 Recombinant Rabbit Monoclonal Antibody (23GB2580), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553557
This item is not returnable.
View return policy
Description
The protein encoded by this gene contains several WD-40 repeats and is found mostly in the nucleus. The expression and localization of this protein are cell cycle dependent. Expression of this gene is upregulated in pancreatic adenocarcinoma. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| PWP1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PWP1 | |
| 2310058A11Rik; 2610205J09Rik; AI098170; AL022879; AV083409; AW550690; C20orf152; endonuclein; IEF-SSP-9502; Keratinocyte protein IEF SSP 9502; nuclear phosphoprotein similar to S. cerevisiae PWP1; periodic tryptophan protein 1 homolog; Pwp1; PWP1 homolog (S. cerevisiae); PWP1 homolog, endonuclein; RGD1310705 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 103136, 11137 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2580 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13610, Q99LL5 | |
| PWP1 | |
| A synthesized peptide derived from human PWP1 (460-501AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |