missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PWP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565221
This item is not returnable.
View return policy
Description
Immunogen sequence: MSLGKPAASLA VHTDKVQTLQ FHPFEAQTLI SGSYDKSVAL YDCRSPDESH RMWRFSGQIE RVTWNHFSPC HFLASTDDGF VYNLDARSDK P Highest antigen sequence identity to the following orthologs - mouse 87%, rat 89%.
The protein encoded by this gene contains several WD-40 repeats and is found mostly in the nucleus. The expression and localization of this protein are cell cycle dependent. Expression of this gene is upregulated in pancreatic adenocarcinoma. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| PWP1 | |
| Polyclonal | |
| Unconjugated | |
| PWP1 | |
| 2310058A11Rik; 2610205J09Rik; AI098170; AL022879; AV083409; AW550690; C20orf152; endonuclein; IEF-SSP-9502; Keratinocyte protein IEF SSP 9502; nuclear phosphoprotein similar to S. cerevisiae PWP1; periodic tryptophan protein 1 homolog; Pwp1; PWP1 homolog (S. cerevisiae); PWP1 homolog, endonuclein; RGD1310705 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11137 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13610 | |
| PWP1 | |
| Recombinant Human PWP1. Recombinant protein control fragment (Product #RP-104694). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction