missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PWP1, Mouse anti-Human, Clone: 3A11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00011137-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MNRSRQVTCVAWVRCGVAKETPDKVELSKEEVKRLIAEAKEKLQEEGGGSDEEETGSPSEDGMQSARTQARPREPLEDGDPEDDRTLDDDELAEYDLDKYDEEGDPDAETLGESLLGLTVYGSNDQDPYVTLKDTEQYEREDFLIKPSDNLIVCGRAEQDQCNLDVHVYNQEEDSFYVHHDILLSAYPLSVEWLNFDPSPDDSTGNYIAVGNMTPVIEVWDLDIVDSLEPVFTLGSKLSKKKKKKGKKSSSAEGHTDAVLDLSWNKLIRNVLASASADNTVILWDMSLGKPAASLAVHTDKVQTLQFHPFEAQTLISGSYDKSVALYDCRSPDESHRMWRFSGQIERVTWNHFSPCHFLASTDDGFVYNLDARSDKPIFTLNAHNDEISGLDLSSQIKGCLVTASADKYVKIWDILGDRPSLVHSRDMKMGVLFCSSCCPDLPFIYALRGQKEGLRVWDISTVSSVNEAFGRRERLVLGSARNSSISGPFGSRSSDTPMESSpecifications
| PWP1 | |
| Monoclonal | |
| Unconjugated | |
| PWP1 homolog (S. cerevisiae) | |
| IEF-SSP-9502 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3A11 | |
| In 1x PBS, pH 7.4 | |
| BC001652 | |
| PWP1 | |
| PWP1 (AAH01652, 1 a.a. ∼ 501 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 11137 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction