Learn More
Ismatec™ PVC Solvent Ready Microbore Extension Tubing
Description
- Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Regulatory approvals to match your unique application
Specifications
Specifications
| Diameter (Metric) Inner | 0.89 mm |
| Material | PVC (Solvent-ready) |
| Diameter (Metric) Outer | 2.82 mm |
| Min. Operating Temperature (Metric) | -50°C |
| Packaging Quantity | 1 Roll |
| Tubing Size | 26 |
| Max. Operating Temperature (Metric) | 74°C |
| Product Type | Extension Pump Tubing |