Learn More
PUS7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554983
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PGFDVIYPKH KIQEAYREML TADNLDIDNM RHKIRDYSLS GAYRKIIIRP QNVSWEVVAY DDPKIPLFNT DVDNLEGKTP PVFASEGKYR ALKMDFSLPP STYATMAIRE VLKMDTSIKN QTQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
PUS7 Polyclonal antibody specifically detects PUS7 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
PUS7 | |
Polyclonal | |
Unconjugated | |
PUS7 | |
C330017I15Rik; FLJ20485; Kiaa1897; pseudouridine synthase 7; pseudouridylate synthase 7; pseudouridylate synthase 7 (putative); pseudouridylate synthase 7 homolog; pseudouridylate synthase 7 homolog (S. cerevisiae); PUS7; RGD1307054 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
296751, 54517, 78697 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PZ0 | |
PUS7 | |
Recombinant protein corresponding to Human PUS7 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only