Learn More
Brand™ Pursept- AF Surface Disinfecting Detergents
Product Code. 15493510 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
2 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15493510 2 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15493510 Supplier Brand™ Supplier No. 44850
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Surface Disinfecting Detergent
Form Liquid
pH 9.5
Appearance Clear Green
Certifications/Compliance DGHM/VAH Tested and RKI Compliant
Composition 100g Pursept-AF Contain 12.5g Didecyl-Dimethyl-Ammonium chloride, 10.4g Alkylpropylenediaminebis-Gua
Container Type Bottle
For Use With (Application) For Use In The Foods Sector
Quantity 1 Ea.
Volume (Metric) 2 L
compliance-icons compliance-icons compliance-icons
Product Identifier
  • pursept AF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Guanidine, N, N'''-1, 3- propanediylbis-, N-coco alkyl derivatives, diacetates,Didecyldimethylammon