Learn More
Invitrogen™ PureLink™ Expi Endotoxin-Free Mega Plasmid Purification Kit
Product Code. 15697878
Click to view available options
Quantity:
4 preps
Unit Size:
Pack of 4
For Research Use Only. All usage must comply with product instructions.
Product Code. 15697878
Brand: Invitrogen™ A31232
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
The PureLink™ Expi Endotoxin-Free Mega Plasmid Purification Kit enables quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange resin.
The PureLink™ Expi Endotoxin-Free Mega Plasmid Purification Kit enables quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange resin. Purify up to 5 mg of high quality plasmid DNA that is suitable for advanced transfection applications such as transfection of sensitive cells lines. The kit utilizes a novel vacuum-assisted purification workflow that dramatically reduces purification time to approximately 2 hours.
  • Simple and fast protocol—unlike other anion exchange-based endotoxin-free plasmid purification systems, a vacuum filtration method is used for lysate clarification and plasmid DNA purification. There is no waiting for gravity-flow columns, and purification typically takes 2 hours.
  • High yield—isolate up to 5 mg of high quality, endotoxin-free plasmid DNA
  • Endotoxin-free plasmid—produces endotoxin-free (less than 0.1 EU/μg) plasmid that is ideal for sensitive applications such as transfection of primary cells and research on gene therapy or plasmid vaccines
  • Purity—ultralow levels of protein, RNA, and genomic DNA contamination
  • Compatibility—purified plasmid is compatible with the Gibco™ ExpiCHO™ and Expi293™ transient mammalian expression systems
  • Endotoxin-free components—includes additional endotoxin-free elution buffer, water, and plasmid storage tubes
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage
• Resuspension Buffer
• Lysis Indicator
• RNase A
• Lysis Buffer
• Precipitation Buffer
• Endotoxin Removal Buffer
• Wash Buffer
• Elution Buffer
• TE Buffer
• Expi Mega Filtration Column
• Expi Mega DNA Binding Column
Endotoxin-free components: elution buffer, water, plasmid storage tubes
Format Spin Column
Isolation Technology Anion Exchange Resin
Sample Type Bacterial Culture
Expression System Mammalian
For Use With (Application) Cloning, PCR
High-throughput Compatibility Manual Protocols
Prep Scale Large Volume Microbial Samples (> 10 mL)
Product Line PureLink
Product Type Plasmid Purification Kit
Quantity 4 preps
Shipping Condition Room Temperature
Target Endotoxin-free Plasmid
Test Time 2 hr.