Learn More
PUMA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55120-1-AP-20UL
 View more versions of this product
Product Code. 16896575
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
55120-1-AP can recognize PUMA alpha 21-24 kDa and PUMA beta 15-18 kDa.
PUMA is one of the pro-apoptotic Bcl-2 family members including Bax and Noxa, which are also transcriptional targets of p53. The PUMA gene encodes two BH3 domain-containing proteins termed PUMA-a and PUMA-b. PUMA proteins bind Bcl-2, localize to the mitochondria, and induce cytochrome c release and apoptosis in response to p53. PUMA may be a direct mediator of p53-induced apoptosis. Antisense inhibition of PUMA expression reduced the apoptotic response to p53, and PUMA is likely to play a role in mediating p53-induced cell death through the cytochrome c/Apaf-1-dependent pathway.
Specifications
Specifications
PUMA
Polyclonal
Unconjugated
BBC3
BBC3, JFY 1, JFY1, PUMA
Rabbit
Antigen Affinity Chromatography
RUO
170770, 27113, 317673
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.3 mg/mL
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80ZG6, Q99ML1, Q9BXH1
Bbc3
Peptide
20 μL
Primary
Rat, Mouse, Human