Learn More
PUM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11586-1-AP-150UL
 View more versions of this product
Product Code. 16868763
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
Specifications
Specifications
PUM2
Polyclonal
Unconjugated
PUM2
KIAA0235, PUM2, PUMH2, Pumilio 2, Pumilio homolog 2, pumilio homolog 2 (Drosophila), PUML2
Rabbit
Antigen Affinity Chromatography
RUO
23369, 298874, 80913
-20°C
Liquid
Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q80U58, Q8TB72
Pum2
PUM2 Fusion Protein Ag2179
150 μL
Primary
Human, Mouse, Rat