missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ PU 23
Description
PU 23 is a multidrug resistance protein (MRP) 4 inhibitor. Sensitizes MRP4-overexpressing cells to the anticancer agent 6-Mercaptopurine (Cat. No. 4103; 6-MP) (IC50 values are 14.11 and 6.06 μM in the absence and presence of compound, respectively). Increases accumulation of 6-MP in MRP4-overexpressing cells.
Specifications
Specifications
| Quantity | 50 mg |
| Molecular Formula | C21H19N3O3S2 |
| Formula Weight | 425.52 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?